Assay Details
| Gene Symbol: Naaladl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009546.1 | NP_001009546.1 | 15 - 16 | 1869 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Naaladl1
Genomic Map