Assay Details
| Gene Symbol: Frmd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026169.4 | NP_080445.1 | 5 - 6 | 527 | 86 | |
| GenBank mRNA | AK004603.2 | - | 5 - 6 | 550 | 86 | |
| AK049224.1 | - | 5 - 6 | 542 | 86 | ||
| AK049233.1 | - | 5 - 6 | 542 | 86 | ||
| AK083902.1 | - | 5 - 6 | 529 | 86 | ||
| AK088639.1 | - | 5 - 6 | 539 | 86 | ||
| AK148434.1 | - | 539 | 86 | |||
| AK168669.1 | - | 5 - 6 | 538 | 85 | ||
| BC004765.1 | - | 5 - 6 | 595 | 3156148 | 86 | |
More Information
Set Membership: |
|

Frmd8
Genomic Map