Assay Details
| Gene Symbol: Pola2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164057.1 | NP_001157529.1 | 9 - 10 | 1264 | 72 | |
| NM_008893.3 | NP_032919.2 | 9 - 10 | 1264 | 72 | ||
| GenBank mRNA | AK087989.1 | - | 9 - 10 | 1260 | 72 | |
| AK133832.1 | - | 9 - 10 | 1250 | 72 | ||
| AK150099.1 | - | 9 - 10 | 1206 | 72 | ||
| AK152974.1 | - | 9 - 10 | 1229 | 72 | ||
| AK153081.1 | - | 9 - 10 | 1231 | 72 | ||
| AK211272.1 | - | 3 - 4 | 321 | 72 | ||
| BC021424.1 | - | 9 - 10 | 1239 | 5011204 | 72 | |
| BC023688.1 | - | 9 - 10 | 1278 | 5343583 | 72 | |
| BC064795.1 | - | 9 - 10 | 1180 | 30111287 | 72 | |
| D13546.1 | - | 9 - 10 | 1197 | 72 | ||
More Information
Set Membership: |
|

Pola2
Genomic Map