Assay Details
| Gene Symbol: Syvn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164709.1 | NP_001158181.1 | 10 - 11 | 1143 | 74 | |
| NM_028769.5 | NP_083045.4 | 10 - 11 | 1510 | 74 | ||
| GenBank mRNA | AK004688.2 | - | 10 - 11 | 997 | 74 | |
| AK122558.1 | - | 8 - 9 | 1629 | 74 | ||
| AK143720.1 | - | 10 - 11 | 1022 | 74 | ||
| AK172097.1 | - | 7 - 8 | 1720 | 74 | ||
| BC042199.1 | - | 10 - 11 | 1005 | 4953165 | 74 | |
| BC046829.1 | - | 10 - 11 | 1143 | 6811574 | 74 | |
| BC057917.1 | - | 10 - 11 | 1424 | 5136398 | 74 | |
| BC080722.1 | - | 10 - 11 | 1028 | 5323695 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Syvn1
Genomic Map