Assay Details
| Gene Symbol: Banf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011793.3 | NP_035923.1 | 1 - 2 | 311 | 65 | |
| GenBank mRNA | AB025349.1 | - | 1 - 2 | 43 | 65 | |
| AK003025.1 | - | 1 - 2 | 87 | 65 | ||
| AK004212.1 | - | 1 - 2 | 70 | 65 | ||
| AK011079.1 | - | 1 - 2 | 92 | 65 | ||
| AK011100.1 | - | 1 - 2 | 90 | 65 | ||
| AK012588.1 | - | 1 - 2 | 73 | 65 | ||
| AK088896.1 | - | 1 - 2 | 67 | 65 | ||
| AK168039.1 | - | 1 - 2 | 93 | 65 | ||
| BC003709.1 | - | 1 - 2 | 80 | 3599828 | 65 | |
| CK780850.1 | - | 1 - 2 | 75 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB025349.1, AK003025.1, AK004212.1, AK011079.1, AK011100.1, AK012588.1, AK088896.1, BC003709.1, NM_011793.3 transcripts.
Set Membership: |
|

Banf1
Genomic Map