Assay Details
| Gene Symbol: Pclo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110796.2 | NP_001104266.1 | 3 - 4 | 3418 | 154 | |
| NM_011995.4 | NP_036125.4 | 3 - 4 | 3418 | 154 | ||
| GenBank mRNA | AB083477.1 | - | 3 - 4 | 3343 | 154 | |
| AB083478.1 | - | 3 - 4 | 3343 | 154 | ||
| Y19185.2 | - | 4 - 5 | 3328 | 154 | ||
| Y19186.2 | - | 4 - 5 | 3328 | 154 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pclo
Genomic Map