| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500000813.mm.1 |
TC0500000093.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500000813.mm.1 |
NM_011348.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500000813.mm.1 |
NM_011348.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500000811.mm.1 |
TC0500000093.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500000811.mm.1 |
NM_011348.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500000811.mm.1 |
NM_011348.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500000812.mm.1 |
TC0500000093.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500000812.mm.1 |
NM_011348.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500000812.mm.1 |
NM_011348.2 |
Refseq |