Assay Details
| Gene Symbol: Sema3e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011348.2 | NP_035478.2 | 2 - 3 | 911 | 92 | |
| GenBank mRNA | AF034744.1 | - | 2 - 3 | 646 | 92 | |
| AK141295.1 | - | 2 - 3 | 913 | 92 | ||
| AK142340.1 | - | 2 - 3 | 738 | 92 | ||
| AK159214.1 | - | 2 - 3 | 702 | 92 | ||
| BC057956.1 | - | 2 - 3 | 809 | 5357516 | 92 | |
| Z80941.1 | - | 2 - 3 | 277 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sema3e
Genomic Map