| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100017307.mm.1 |
NM_001160235.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100017307.mm.1 |
NM_001160235.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100017307.mm.1 |
NM_001160236.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100017307.mm.1 |
NM_001160236.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100017307.mm.1 |
NM_174997.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100017307.mm.1 |
NM_174997.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100017307.mm.1 |
TC0100002163.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100017305.mm.1 |
NM_001160235.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100017305.mm.1 |
NM_001160235.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100017305.mm.1 |
NM_001160236.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100017305.mm.1 |
NM_001160236.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100017305.mm.1 |
NM_174997.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100017305.mm.1 |
NM_174997.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100017305.mm.1 |
TC0100002163.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100017306.mm.1 |
NM_001160235.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100017306.mm.1 |
NM_001160235.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100017306.mm.1 |
NM_001160236.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100017306.mm.1 |
NM_001160236.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100017306.mm.1 |
NM_174997.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100017306.mm.1 |
NM_174997.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100017306.mm.1 |
TC0100002163.mm.1 |
MTA TC |