Assay Details
| Gene Symbol: Fam168b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160235.1 | NP_001153707.1 | 5 - 6 | 566 | 86 | |
| NM_174997.2 | NP_778162.1 | 4 - 5 | 530 | 86 | ||
| GenBank mRNA | AK144298.1 | - | 4 - 5 | 430 | 86 | |
| AK220453.1 | - | 4 - 5 | 530 | 86 | ||
| BC040511.1 | - | 4 - 5 | 507 | 4977517 | 86 | |
| BC043098.1 | - | 4 - 5 | 454 | 6406013 | 86 | |
| BC059247.1 | - | 4 - 5 | 507 | 6409420 | 86 | |
| CF538379.1 | - | 5 - 6 | 527 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam168b
Genomic Map