Assay Details
| Gene Symbol: Cnot2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC063105.1 | - | 3 - 4 | 343 | 5003026 | 125 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC063105.1 transcripts.
Set Membership: |
|

Cnot2
Genomic Map