Assay Details
| Gene Symbol: Syn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013722.3 | NP_038750.2 | 9 - 10 | 1242 | 68 | |
| GenBank mRNA | AF192754.1 | - | 9 - 10 | 1187 | 68 | |
| AF498252.1 | - | 8 - 9 | 989 | 68 | ||
| AF498253.1 | - | 8 - 9 | 989 | 68 | ||
| AK080557.1 | - | 9 - 10 | 1204 | 68 | ||
| BC107397.2 | - | 8 - 9 | 1108 | 40058225 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Syn3
Genomic Map