Assay Details
| Gene Symbol: 4930555G07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016129.1 | - | 1 - 2 | 227 | 63 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1700113H08 gene | 1700113H08Rik | -161.0 | Sense | Chr. 10 - 87040467 - 87230599 on NCBI Build GRCm38 | |
| Downstream | achaete-scute complex homolog 1 (Drosophila) | Ascl1 |
AI225900, ASH1, Mash1, bHLHa46 |
255.3kb | Anti-sense | Chr. 10 - 87491041 - 87493660 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

4930555G07Rik
Genomic Map