| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
134990 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
680016 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
775665 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
861961 |
AK029058.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
861961 |
AK137781.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
861961 |
BC021511.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
861961 |
XM_001479246.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
861961 |
XM_916928.3 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
272666 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
176149 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P155512 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P155512 |
- |
- |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P118040 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P155512 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08366943 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27318700 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27318683 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08769086 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08366964 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27318651 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27318640 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27318661 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27318670 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27318650 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27318672 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27318667 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08755726 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27318674 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27318676 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27318646 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27318680 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27318677 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27318647 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08955185 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27318687 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27318691 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27318665 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08366956 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08366968 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27318654 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27318704 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27318658 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27318678 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27318702 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27318690 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27318660 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08366966 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27318656 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08572301 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27318668 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27318638 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27318642 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27318663 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27318679 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27318692 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27318696 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27318685 |
- |
- |