Assay Details
| Gene Symbol: Lmbrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026719.2 | NP_080995.2 | 12 - 13 | 1380 | 74 | |
| GenBank mRNA | AK032560.1 | - | 1679 | 74 | ||
| AK158982.1 | - | 12 - 13 | 1216 | 74 | ||
| AK167080.1 | - | 12 - 13 | 1199 | 74 | ||
| AY061989.1 | - | 11 - 12 | 1091 | 74 | ||
| BC025579.1 | - | 12 - 13 | 1212 | 5325113 | 74 | |
| BC027774.1 | - | 12 - 13 | 1263 | 5346846 | 74 | |
| BC031902.1 | - | 12 - 13 | 1203 | 5362166 | 74 | |
| BC039053.1 | - | 12 - 13 | 1294 | 2655321 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lmbrd1
Genomic Map