Assay Details
| Gene Symbol: Lmbrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026719.2 | NP_080995.2 | 13 - 14 | 1536 | 88 | |
| GenBank mRNA | AK032560.1 | - | 1835 | 88 | ||
| AK152170.1 | - | 13 - 14 | 1347 | 88 | ||
| AK153237.1 | - | 13 - 14 | 1347 | 88 | ||
| AK158982.1 | - | 13 - 14 | 1372 | 88 | ||
| AK167080.1 | - | 13 - 14 | 1355 | 88 | ||
| AY061989.1 | - | 12 - 13 | 1247 | 88 | ||
| BC025579.1 | - | 13 - 14 | 1368 | 5325113 | 88 | |
| BC027774.1 | - | 13 - 14 | 1419 | 5346846 | 88 | |
| BC031902.1 | - | 13 - 14 | 1359 | 5362166 | 88 | |
| BC039053.1 | - | 13 - 14 | 1450 | 2655321 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lmbrd1
Genomic Map