Assay Details
| Gene Symbol: Lmbrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026719.2 | NP_080995.2 | 14 - 15 | 1618 | 91 | |
| GenBank mRNA | AK003084.1 | - | 1 - 2 | 67 | 91 | |
| AK032560.1 | - | 1917 | 91 | |||
| AK152170.1 | - | 14 - 15 | 1429 | 91 | ||
| AK153237.1 | - | 14 - 15 | 1429 | 91 | ||
| AK158982.1 | - | 14 - 15 | 1454 | 91 | ||
| AK167080.1 | - | 14 - 15 | 1437 | 91 | ||
| AY061989.1 | - | 13 - 14 | 1329 | 91 | ||
| BC025579.1 | - | 14 - 15 | 1450 | 5325113 | 91 | |
| BC027774.1 | - | 14 - 15 | 1501 | 5346846 | 91 | |
| BC031902.1 | - | 14 - 15 | 1441 | 5362166 | 91 | |
| BC039053.1 | - | 14 - 15 | 1532 | 2655321 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lmbrd1
Genomic Map