Assay Details
| Gene Symbol: Lmbrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026719.2 | NP_080995.2 | 3 - 4 | 504 | 91 | |
| GenBank mRNA | AK032560.1 | - | 803 | 91 | ||
| AK152170.1 | - | 3 - 4 | 315 | 91 | ||
| AK153237.1 | - | 3 - 4 | 315 | 91 | ||
| AK158982.1 | - | 3 - 4 | 340 | 91 | ||
| AK167080.1 | - | 3 - 4 | 323 | 91 | ||
| AY061989.1 | - | 2 - 3 | 215 | 91 | ||
| BC025579.1 | - | 3 - 4 | 336 | 5325113 | 91 | |
| BC027774.1 | - | 3 - 4 | 387 | 5346846 | 91 | |
| BC031902.1 | - | 3 - 4 | 327 | 5362166 | 91 | |
| BC039053.1 | - | 3 - 4 | 418 | 2655321 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lmbrd1
Genomic Map