Assay Details
| Gene Symbol: Nap1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK007322.1 | - | 13 - 14 | 1228 | 99 | |
| AK032896.1 | - | 13 - 14 | 1236 | 99 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nap1l1| Gene Symbol: Nap1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK007322.1 | - | 13 - 14 | 1228 | 99 | |
| AK032896.1 | - | 13 - 14 | 1236 | 99 | ||
Set Membership: |
|