Assay Details
| Gene Symbol: Il1rl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025602.3 | NP_001020773.1 | 9 - 10 | 1482 | 102 | |
| GenBank mRNA | D13695.1 | - | 8 - 9 | 1174 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Il1rl1
Genomic Map