Assay Details
| Gene Symbol: Il18r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161842.1 | NP_001155314.1 | 5 - 6 | 921 | 151 | |
| NM_001161843.1 | NP_001155315.1 | 5 - 6 | 793 | 151 | ||
| NM_008365.2 | NP_032391.1 | 4 - 5 | 846 | 151 | ||
| GenBank mRNA | AK087129.1 | - | 4 - 5 | 795 | 151 | |
| AK089237.1 | - | 5 - 6 | 922 | 151 | ||
| AK170705.1 | - | 5 - 6 | 882 | 151 | ||
| BC020296.1 | - | 4 - 5 | 608 | 151 | ||
| BC023240.1 | - | 5 - 6 | 762 | 4483261 | 151 | |
| U43673.1 | - | 4 - 5 | 846 | 151 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il18r1
Genomic Map