Assay Details
| Gene Symbol: Il18r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161842.1 | NP_001155314.1 | 8 - 9 | 1255 | 131 | |
| NM_001161843.1 | NP_001155315.1 | 8 - 9 | 1127 | 131 | ||
| NM_008365.2 | NP_032391.1 | 7 - 8 | 1180 | 131 | ||
| GenBank mRNA | AK089237.1 | - | 8 - 9 | 1256 | 131 | |
| BC020296.1 | - | 7 - 8 | 942 | 131 | ||
| BC023240.1 | - | 8 - 9 | 1096 | 4483261 | 131 | |
| U43673.1 | - | 7 - 8 | 1180 | 131 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il18r1
Genomic Map