Assay Details
| Gene Symbol: Slc6a15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252330.1 | NP_001239259.1 | 1 - 2 | 348 | 133 | |
| GenBank mRNA | AK036136.1 | - | 1 - 2 | 327 | 133 | |
| AY149281.1 | - | 1 - 2 | 323 | 133 | ||
| BC076593.1 | - | 1 - 2 | 348 | 30652984 | 133 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK036136.1, AY149281.1, BC076593.1, NM_001252330.1 transcripts.
Set Membership: |
|

Slc6a15
Genomic Map