Assay Details
| Gene Symbol: Acss3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142804.1 | NP_001136276.1 | 11 - 12 | 1572 | 157 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acss3| Gene Symbol: Acss3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142804.1 | NP_001136276.1 | 11 - 12 | 1572 | 157 | |
Set Membership: |
|