Assay Details
| Gene Symbol: Acss3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142804.1 | NP_001136276.1 | 3 - 4 | 716 | 90 | |
| NM_198636.3 | NP_941038.2 | 3 - 4 | 716 | 90 | ||
| GenBank mRNA | AK033376.1 | - | 3 - 4 | 721 | 90 | |
| BC113198.1 | - | 3 - 4 | 680 | 40062399 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acss3
Genomic Map