| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000021352.mm.1 |
NM_198636.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000021352.mm.1 |
NM_198636.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000021352.mm.1 |
TC1000002814.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000021353.mm.1 |
NM_198636.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000021353.mm.1 |
NM_198636.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000021353.mm.1 |
TC1000002814.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000021355.mm.1 |
NM_001142804.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000021355.mm.1 |
NM_001142804.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000021355.mm.1 |
NM_198636.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000021355.mm.1 |
NM_198636.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000021355.mm.1 |
TC1000002814.mm.1 |
MTA TC |