Assay Details
| Gene Symbol: Mettl25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207522.2 | NP_997405.2 | 8 - 9 | 1524 | 116 | |
| GenBank mRNA | AK144620.1 | - | 8 - 9 | 1516 | 116 | |
| AK148535.1 | - | 9 - 10 | 1701 | 116 | ||
| BC067068.1 | - | 8 - 9 | 1498 | 30606425 | 116 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mettl25
Genomic Map