Assay Details
| Gene Symbol: 4921524L21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC049627.1 | - | 2 - 3 | 155 | 6706381 | 133 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC049627.1 transcripts.
Set Membership: |
|

4921524L21Rik
Genomic Map