Assay Details
| Gene Symbol: Ehbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252515.1 | NP_001239444.1 | 15 - 16 | 2973 | 110 | |
| NM_153078.4 | NP_694718.4 | 15 - 16 | 2898 | 110 | ||
| GenBank mRNA | AF424697.1 | - | 14 - 15 | 2748 | 110 | |
| AK173055.1 | - | 15 - 16 | 2974 | 110 | ||
| BC037023.1 | - | 2 - 3 | 525 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ehbp1
Genomic Map