Assay Details
Gene Symbol: 4933406G16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_046037.1 | 2 - 3 | 943 | 73 | ||
GenBank mRNA | AK016692.1 | - | 2 - 3 | 944 | 73 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Meis homeobox 1 | Meis1 |
C530044H18Rik, Evi8 |
-46.4kb | Anti-sense | Chr. 11 - 18880428 - 19020482 on NCBI Build GRCm38 |
Downstream |
sprouty-related, EVH1 domain containing 2 |
Spred2 | C79158 | 855.8kb | Sense | Chr. 11 - 19924442 - 20024026 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
