Assay Details
| Gene Symbol: Taf9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001176.2 | NP_001001176.2 | 6 - 7 | 796 | 70 | |
| NM_001167988.1 | NP_001161460.1 | 7 - 8 | 734 | 70 | ||
| GenBank mRNA | AK131710.1 | - | 7 - 8 | 735 | 70 | |
| AK135097.1 | - | 6 - 7 | 70 | |||
| BC051635.1 | - | 6 - 7 | 594 | 70 | ||
| BC066223.1 | - | 6 - 7 | 587 | 30468965 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
DNA-templated transcription, initiation
regulation of transcription, DNA-templated
positive regulation of cell growth
negative regulation of apoptotic process
protein stabilization
molecular_function
protein heterodimerization activity
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
DNA-templated transcription, initiation
regulation of transcription, DNA-templated
positive regulation of cell growth
negative regulation of apoptotic process
protein stabilization
molecular_function
protein heterodimerization activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32583453 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32583458 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32583454 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32583450 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32583451 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32583456 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32583457 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2244722 | BC066223.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2244722 | AK135097.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2244722 | BC051635.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2244722 | NM_001001176.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2244722 | NM_001167988.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 413750 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 489097 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 545044 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 505670 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 633846 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 761806 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 160062 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 88624 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 162144 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136684 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P101701 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2244722 | BC066223.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2244722 | AK135097.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2244722 | BC051635.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2244722 | NM_001001176.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2244722 | NM_001167988.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P176634 | BC066223.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P176634 | AK135097.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P176634 | BC051635.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P176634 | NM_001001176.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P176634 | NM_001167988.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32583451 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32583462 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32583446 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32583453 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32583463 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32583449 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32583476 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32583465 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32583459 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32583460 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32583448 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32583454 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32583439 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32583464 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32583482 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32583467 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32583452 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32583458 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32583455 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32583475 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32583473 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32583469 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32583468 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32583450 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32583471 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32583479 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32583457 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32583445 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32583443 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32583477 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32583480 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32583442 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32583456 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32583437 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136684 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P101701 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09385188 | - | - |
| Whole Mouse Genome G4122A | A_52_P176634 | BC066223.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P176634 | AK135097.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P176634 | BC051635.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P176634 | NM_001001176.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P176634 | NM_001167988.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P592749 | BC066223.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P592749 | CA559941.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P592749 | AK135097.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P592749 | BC051635.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P592749 | NM_001001176.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104773 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136684 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P101701 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14517497 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14991296 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14517486 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32583454 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32583450 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32583458 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32583457 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32583451 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32583453 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32583456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32583482 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583425 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583437 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583473 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583455 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583463 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583417 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583414 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583415 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583426 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583457 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583412 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14991300 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583411 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583419 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583443 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583429 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14552306 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583465 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583442 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583434 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583427 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14991296 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583464 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14755018 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583462 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583490 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583477 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583431 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14639752 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583467 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583423 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583471 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583422 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14845579 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583449 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14991298 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583430 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583480 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14803478 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583439 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583459 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583491 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583488 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583433 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583448 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583445 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583446 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14779840 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583485 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583482 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583440 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583469 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14517486 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15057962 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583421 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583483 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14517497 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32583420 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10606322 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10606318 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10606319 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10606321 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10606320 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10606317 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10606316 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434960_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1447829_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10606315 | - | - |
| Mouse Genome 430 2.0 Array | 1447829_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1434960_at | - | - |
| Mouse Genome 430 2.0 Array | 1447829_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1434960_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 407786_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 407786_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00017949.mm.1 | NM_001001176.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017949.mm.1 | NM_001001176.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017949.mm.1 | NM_001167988.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017949.mm.1 | NM_001167988.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00017949.mm.1 | TC0X00002826.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1447829_x_at | - | - |
| Mouse Expression Set 430 | 1434960_at | - | - |
| Mouse Expression Set 430 | 1434960_PM_at | - | - |
| Mouse Expression Set 430 | 1447829_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1447829_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434960_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434960_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1447829_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 724112 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1243080 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2859348 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2859348 | - | - |

Taf9b
Genomic Map