Assay Details
| Gene Symbol: Mdh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008618.3 | NP_032644.3 | 5 - 6 | 700 | 107 | |
| GenBank mRNA | AK002404.1 | - | 5 - 6 | 558 | 107 | |
| AK005237.1 | - | 5 - 6 | 595 | 107 | ||
| AK008270.1 | - | 5 - 6 | 572 | 107 | ||
| AK164785.1 | - | 5 - 6 | 581 | 107 | ||
| AK168545.1 | - | 5 - 6 | 538 | 107 | ||
| BC050940.1 | - | 5 - 6 | 566 | 6820601 | 107 | |
| M29462.1 | - | 5 - 6 | 579 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mdh1
Genomic Map