Assay Details
| Gene Symbol: 4930481A15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027938.2 | 1 - 2 | 265 | 60 | ||
| GenBank mRNA | BI202508.1 | - | 2 - 3 | 60 | ||
| BM937611.1 | - | 1 - 2 | 194 | 60 | ||
More Information
Set Membership: |
|

4930481A15Rik
Genomic Map