Assay Details
| Gene Symbol: Syvn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164709.1 | NP_001158181.1 | 9 - 10 | 1028 | 100 | |
| NM_028769.5 | NP_083045.4 | 9 - 10 | 1395 | 100 | ||
| GenBank mRNA | AK004688.2 | - | 9 - 10 | 882 | 100 | |
| AK122558.1 | - | 7 - 8 | 1514 | 100 | ||
| AK143720.1 | - | 9 - 10 | 907 | 100 | ||
| AK172097.1 | - | 6 - 7 | 1605 | 100 | ||
| BC042199.1 | - | 9 - 10 | 890 | 4953165 | 100 | |
| BC046829.1 | - | 9 - 10 | 1028 | 6811574 | 100 | |
| BC057917.1 | - | 9 - 10 | 1309 | 5136398 | 100 | |
| BC080722.1 | - | 9 - 10 | 913 | 5323695 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Syvn1
Genomic Map