Assay Details
| Gene Symbol: Men1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168490.1 | NP_001161962.1 | 7 - 8 | 1606 | 78 | |
| GenBank mRNA | BC036287.1 | - | 7 - 8 | 1534 | 4189611 | 78 |
More Information
Set Membership: |
|

Men1
Genomic Map