Assay Details
| Gene Symbol: Esyt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028731.5 | NP_083007.2 | 2 - 3 | 496 | 103 | |
| GenBank mRNA | AK044360.1 | - | 2 - 3 | 497 | 103 | |
| AK157352.1 | - | 2 - 3 | 465 | 103 | ||
| BC138937.1 | - | 2 - 3 | 405 | 8861959 | 103 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK044360.1 transcripts.
Set Membership: |
|

Esyt2
Genomic Map