Assay Details
| Gene Symbol: Esyt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028731.5 | NP_083007.2 | 6 - 7 | 862 | 76 | |
| GenBank mRNA | AK157352.1 | - | 6 - 7 | 831 | 76 | |
| BC138937.1 | - | 6 - 7 | 771 | 8861959 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Esyt2
Genomic Map