Assay Details
| Gene Symbol: Gm16152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK040668.1 | - | 2 - 3 | 199 | 72 | |
| AK048050.1 | - | 312 | 72 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
AF4/FMR2 family, member 3 |
Aff3 |
3222402O04Rik, A730046J16, LAF-4, Laf4 |
Anti-sense | Chr. 1 - 38175991 - 38627837 on NCBI Build GRCm38 | |
| Downstream | LON peptidase N-terminal domain and ring finger 2 | Lonrf2 |
2900060P06Rik, AI851349 |
403.7kb | Anti-sense | Chr. 1 - 38793669 - 38836719 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm16152
Genomic Map