Assay Details
| Gene Symbol: Smap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290683.1 | NP_001277612.1 | 6 - 7 | 547 | 73 | |
| NM_028534.3 | NP_082810.1 | 6 - 7 | 817 | 73 | ||
| NR_110972.1 | 6 - 7 | 543 | 73 | |||
| GenBank mRNA | AK014966.1 | - | 6 - 7 | 501 | 73 | |
| AK132868.1 | - | 6 - 7 | 544 | 73 | ||
| BC006946.1 | - | 6 - 7 | 725 | 2810817 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smap1
Genomic Map