Assay Details
| Gene Symbol: Smap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290683.1 | NP_001277612.1 | 7 - 8 | 673 | 78 | |
| NM_028534.3 | NP_082810.1 | 7 - 8 | 943 | 78 | ||
| NR_110972.1 | 7 - 8 | 669 | 78 | |||
| GenBank mRNA | AK006817.1 | - | 771 | 78 | ||
| AK014888.1 | - | 7 - 8 | 629 | 78 | ||
| AK014966.1 | - | 7 - 8 | 627 | 78 | ||
| AK132868.1 | - | 7 - 8 | 670 | 78 | ||
| BC006946.1 | - | 7 - 8 | 851 | 2810817 | 78 | |
More Information
Set Membership: |
|

Smap1
Genomic Map