Assay Details
| Gene Symbol: Smap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290683.1 | NP_001277612.1 | 8 - 9 | 834 | 112 | |
| NM_028534.3 | NP_082810.1 | 8 - 9 | 1104 | 112 | ||
| NR_110972.1 | 8 - 9 | 830 | 112 | |||
| GenBank mRNA | AK006817.1 | - | 932 | 112 | ||
| AK014966.1 | - | 8 - 9 | 788 | 111 | ||
| AK132868.1 | - | 8 - 9 | 831 | 112 | ||
| BC006946.1 | - | 8 - 9 | 1012 | 2810817 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smap1
Genomic Map