Assay Details
| Gene Symbol: Camkv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145621.2 | NP_663596.1 | 2 - 3 | 238 | 64 | |
| GenBank mRNA | AK085143.1 | - | 244 | 64 | ||
| AK147323.1 | - | 2 - 3 | 226 | 64 | ||
| BC017634.1 | - | 2 - 3 | 238 | 4527857 | 64 | |
| BC022169.1 | - | 2 - 3 | 233 | 64 | ||
| BC086762.1 | - | 2 - 3 | 222 | 64 | ||
| BC111033.1 | - | 2 - 3 | 233 | 5368225 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Camkv
Genomic Map