Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100001429.mm.1 |
NM_178601.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100001429.mm.1 |
NM_178601.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100001429.mm.1 |
TC0100000222.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100001430.mm.1 |
NM_178601.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100001430.mm.1 |
NM_178601.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100001430.mm.1 |
TC0100000222.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100001427.mm.1 |
NM_178601.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100001427.mm.1 |
NM_178601.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100001427.mm.1 |
TC0100000222.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100001428.mm.1 |
NM_178601.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100001428.mm.1 |
NM_178601.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100001428.mm.1 |
TC0100000222.mm.1 |
MTA TC |