Assay Details
| Gene Symbol: Gm12132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK042535.1 | - | 1 - 2 | 178 | 135 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 4930553C11 gene | 4930553C11Rik | -618.0kb | Anti-sense | Chr. 11 - 39307703 - 39390225 on NCBI Build GRCm38 | |
| Downstream |
methionine adenosyltransferase II, beta |
Mat2b |
1110064C04Rik, 2410018D16Rik, AI182287, AU022853, MAT-II, MATIIbeta, TGR |
669.1kb | Anti-sense | Chr. 11 - 40679314 - 40695203 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm12132
Genomic Map