Assay Details
| Gene Symbol: C330024C12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046016.1 | 2 - 3 | 852 | 102 | ||
| GenBank mRNA | AK082808.1 | - | 2 - 3 | 853 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | suppressor of cytokine signaling 5 | Socs5 |
1810018L08Rik, Cish5, mKIAA0671 |
-55.0kb | Anti-sense | Chr. 17 - 87107407 - 87137647 on NCBI Build GRCm38 |
| Downstream | multiple coagulation factor deficiency 2 | Mcfd2 |
1810021C21Rik, F5f8d, Lman1ip, Sdnsf |
61.3kb | Sense | Chr. 17 - 87254443 - 87265947 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

C330024C12Rik
Genomic Map