Assay Details
| Gene Symbol: Gm14809 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK048032.1 | - | 93 | 73 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 14812 | Gm14812 |
ENSMUSG00000073051, OTTMUSG00000018066 |
Sense | Chr. X - 99405331 - 99443797 on NCBI Build GRCm38 | |
| Downstream |
praja ring finger 1, E3 ubiquitin protein ligase |
Pja1 | praja-1 | 46.8kb | Sense | Chr. X - 99465734 - 99471268 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm14809
Genomic Map