Assay Details
| Gene Symbol: Gm16041 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK044237.1 | - | 1 - 2 | 271 | 108 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | regulator of G-protein signaling 20 | Rgs20 |
2900073E09Rik, Rgsz1 |
Anti-sense | Chr. 1 - 4909573 - 5070308 on NCBI Build GRCm38 | |
| Downstream |
ATPase, H+ transporting, lysosomal V1 subunit H |
Atp6v1h |
0710001F19Rik, AU022349, CGI-11, SFD, SFDalpha, SFDbeta, VMA13 |
111.2kb | Sense | Chr. 1 - 5082978 - 5162549 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm16041
Genomic Map