Assay Details
| Gene Symbol: A930009A15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029982.1 | NP_084258.1 | 4 - 5 | 543 | 65 | |
| GenBank mRNA | AK020834.1 | - | 4 - 5 | 544 | 65 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
zinc finger, C3H1-type containing |
Zfc3h1 |
BC033596, Ccdc131, Psrc2 |
-147.0kb | Sense | Chr. 10 - 115384920 - 115432772 on NCBI Build GRCm38 |
| Downstream | tetraspanin 8 | Tspan8 |
C76990, E330007O21Rik, Tm4sf3 |
234.4kb | Sense | Chr. 10 - 115816817 - 115849893 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

A930009A15Rik
Genomic Map