Assay Details
| Gene Symbol: D230030E09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045947.1 | 1 - 2 | 226 | 97 | ||
| GenBank mRNA | AK084355.1 | - | 1 - 2 | 228 | 97 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | trans-acting transcription factor 4 | Sp4 |
5730497N03Rik, HF-1b, HF1-b |
-228.0kb | Sense | Chr. 12 - 118231686 - 118301440 on NCBI Build GRCm38 |
| Downstream | trans-acting transcription factor 8 | Sp8 |
D930049B17Rik, mBtd |
315.6kb | Anti-sense | Chr. 12 - 118845840 - 118852578 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

D230030E09Rik
Genomic Map