Assay Details
| Gene Symbol: Gm19689 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045094.1 | 1 - 2 | 317 | 66 | ||
| GenBank mRNA | AK051223.1 | - | 1 - 2 | 318 | 66 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
solute carrier family 8 (sodium/calcium exchanger), member 1 |
Slc8a1 |
AI852629, AV344025, D930008O12Rik, Ncx1 |
-1.3mb | Sense | Chr. 17 - 81373105 - 81738387 on NCBI Build GRCm38 |
| Downstream |
protein kinase domain containing, cytoplasmic |
Pkdcc |
AI115348, AW548124, Adtk1, ESTM17, MAd1, Sgk493, Vlk, X83346 |
137.1kb | Anti-sense | Chr. 17 - 83215283 - 83225069 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm19689
Genomic Map